Northern Ireland
Order Entry
Anti-STAT2 Rabbit Polyclonal Antibody
  BIRBORB48683-100
 :  Biorbyt
Anti-STAT2 Rabbit Polyclonal Antibody
Catalog # BIRBORB48683-100
Supplier:  Biorbyt
 
 
   
  1 * 100 µl

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    signal transducer and activator of transcription 2, 113 kDa
  • Antigen symbol:
    STAT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Format:
    ascites fluid
  • Storage buffer:
    0,03% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Peptide
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: STAT2 (signal transducer and activator of transcription 2, 113kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human