Northern Ireland
Order Entry
Anti-ACTB Mouse Monoclonal Antibody
Anti-ACTB Mouse Monoclonal Antibody
  BIRBORB48086-100
 :  Biorbyt
Anti-ACTB Mouse Monoclonal Antibody
  BIRBORB48086-100
 :  Biorbyt
 
 
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Actin beta
  • Antigen symbol:
    ACTB
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Format:
    ascites fluid
  • Storage buffer:
    0,03% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    2,33 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Human Actin fulllength protein
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Western Blot: 1:5000-1:10000; Immunofluorescence: 1:500-1:1000; Immunochemistry: 1:500;

Type: Primary
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: