Northern Ireland
Order Entry
Anti-AMY1A Rabbit Polyclonal Antibody
  BIRBORB47953-100
 :  Biorbyt
undefined
Anti-AMY1A Rabbit Polyclonal Antibody
  BIRBORB47953-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Amylase, Alpha 1A (Salivary)
  • Antigen symbol:
    AMY1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    lyophilized
  • Amino acid number:
    30 to 44
  • Storage buffer:
    5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
  • Sequence:
    HLFEWRWVDIALECE
  • Storage temperature:
    Cool
  • Concentration:
    500 ug/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human AMY1A(30-44aa HLFEWRWVDIALECE), different from the related rat and mouse sequences by one amino acid.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: AMY1A (amylase, alpha 1A (salivary))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: