Northern Ireland
Order Entry
Anti-CD17 Mouse Monoclonal Antibody
  BIRBORB44742-0.1
 :  Biorbyt
Anti-CD17 Mouse Monoclonal Antibody
  BIRBORB44742-0.1
 :  Biorbyt
 
 
   
  1 * 0,1 mg

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD17
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: CD17
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human