Northern Ireland
Order Entry
Anti-EVI2B Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anti-EVI2B Mouse Monoclonal Antibody (PE (Phycoerythrin))
Catalog # BIRBORB44741-100
Supplier:  Biorbyt
Anti-EVI2B Mouse Monoclonal Antibody (PE (Phycoerythrin))
  BIRBORB44741-100
 :  Biorbyt
 
 
   
  1 * 100 Tests

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ecotropic viral integration site 2B
  • Antigen symbol:
    EVI2B
  • Clonality:
    Monoclonal
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 test
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: EVI2B (ecotropic viral integration site 2B)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human