Anti-PRNP Mouse Monoclonal Antibody
BIRBORB44710-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:Prion Protein
- Antigen symbol:PRNP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: PRNP (prion protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human