Anti-CDH1 Mouse Monoclonal Antibody
Catalog # BIRBORB44675-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:cadherin 1, type 1, E-cadherin (epithelial)
- Antigen symbol:CDH1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 test
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human