Anti-ANPEP Mouse Monoclonal Antibody (FITC (Fluorescein))
BIRBORB44584-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Alanyl (Membrane) Aminopeptidase
- Antigen symbol:ANPEP
- Clonality:Monoclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 test
- Pk:100 Tests
Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: