Northern Ireland
Order Entry
Anti-TUBA Mouse Monoclonal Antibody (Biotin)
Anti-TUBA Mouse Monoclonal Antibody (Biotin)
Catalog # BIRBORB44530-0.1
Supplier:  Biorbyt
Anti-TUBA Mouse Monoclonal Antibody (Biotin)
Catalog # BIRBORB44530-0.1
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 0,1 mg

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    tubulin, alpha
  • Antigen symbol:
    TUBA
  • Clonality:
    Monoclonal
  • Conjugation:
    Biotin
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

 

Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: