Anti-CD36 Mouse Monoclonal Antibody (PE (Phycoerythrin))
BIRBORB44518-100
: Biorbyt
- Antibody type:Primary
- Antigen name:CD36 molecule (thrombospondin receptor)
- Antigen symbol:CD36
- Clonality:Monoclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 test
- Pk:100 Tests
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human