Northern Ireland
Order Entry
Anti-CD36 Mouse Monoclonal Antibody (FITC (Fluorescein))
  BIRBORB44517-100
 :  Biorbyt
Anti-CD36 Mouse Monoclonal Antibody (FITC (Fluorescein))
  BIRBORB44517-100
 :  Biorbyt
 
 
   
  1 * 100 Tests

 

  • Antibody type:
    Primary
  • Antigen name:
    CD36 molecule (thrombospondin receptor)
  • Antigen symbol:
    CD36
  • Clonality:
    Monoclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 test
  • Pk:
    100 Tests

 

 

Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human