Northern Ireland
Order Entry
Anti-CD58 Mouse Monoclonal Antibody (FITC (Fluorescein))
Anti-CD58 Mouse Monoclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB44372-100
Supplier:  Biorbyt
Anti-CD58 Mouse Monoclonal Antibody (FITC (Fluorescein))
  BIRBORB44372-100
 :  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 100 Tests

 

  • Antibody type:
    Primary
  • Antigen name:
    CD58 molecule
  • Antigen symbol:
    CD58
  • Clonality:
    Monoclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 test
  • Pk:
    100 Tests

 

About this item

Type: Primary
Antigen: CD58 (CD58 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: