Northern Ireland
Order Entry
Anti-CD53 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anti-CD53 Mouse Monoclonal Antibody (PE (Phycoerythrin))
  BIRBORB44316-100
 :  Biorbyt
Anti-CD53 Mouse Monoclonal Antibody (PE (Phycoerythrin))
  BIRBORB44316-100
 :  Biorbyt
 
 
   
  1 * 100 Tests

 

  • Antibody type:
    Primary
  • Antigen name:
    CD53 molecule
  • Antigen symbol:
    CD53
  • Clonality:
    Monoclonal
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 test
  • Pk:
    100 Tests

 

 

Type: Primary
Antigen: CD53 (CD53 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human