Anti-IGF2R Mouse Monoclonal Antibody
BIRBORB44197-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:Insulin-like Growth Factor 2 Receptor
- Antigen symbol:IGF2R
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: IGF2R (insulin-like growth factor 2 receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: