Anti-KRT19 Mouse Monoclonal Antibody
BIRBORB43666-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:keratin 19
- Antigen symbol:KRT19
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: KRT19 (keratin 19)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human