Anti-ICAM1 Mouse Monoclonal Antibody
Catalog # BIRBORB43582-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:intercellular adhesion molecule 1
- Antigen symbol:ICAM1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 test
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human