Northern Ireland
Order Entry
Anti-IgG1 (Mouse) None Antibody
Anti-IgG1 (Mouse) None Antibody
  BIRBORB43526-0.1
 :  Biorbyt
Anti-IgG1 (Mouse) None Antibody
  BIRBORB43526-0.1
 :  Biorbyt
 
 
   
  1 * 0,1 mg

 

  • Antibody type:
    Control
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage buffer:
    Low Endotoxin.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG1
Reactivity: