Anti-FBL Rabbit Polyclonal Antibody
Catalog # BIRBORB43432-100
Supplier: Biorbyt
- Antibody type:Primary
- Antigen name:Fibrillarin
- Antigen symbol:FBL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µl
Type: Primary
Antigen: FBL (fibrillarin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat