Northern Ireland
Order Entry
Anti-STAT2 Rabbit Polyclonal Antibody
Catalog # BIRBORB43348-100
Supplier:  Biorbyt
undefined
Anti-STAT2 Rabbit Polyclonal Antibody
Catalog # BIRBORB43348-100
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    signal transducer and activator of transcription 2, 113 kDa
  • Antigen symbol:
    STAT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Tyr690
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molecular weight:
    93 - 101 kDa
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human Stat2 around the phosphorylation site of Tyr690.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: STAT2 (signal transducer and activator of transcription 2, 113kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat