Anti-EFNB2 Rabbit Polyclonal Antibody
BIRBORB43340-100
: Biorbyt
- Antibody type:Primary
- Antigen name:ephrin-B2
- Antigen symbol:EFNB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Modification:phospho-Tyr330
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: EFNB2 (ephrin-B2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: