Northern Ireland
Order Entry
Anti-GFP Rabbit Polyclonal Antibody
Anti-GFP Rabbit Polyclonal Antibody
  BIRBORB43335-100
 :  Biorbyt
undefined
Anti-GFP Rabbit Polyclonal Antibody
  BIRBORB43335-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Green Fluorescence Protein
  • Antigen symbol:
    GFP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: