Northern Ireland
Order Entry
Anti-IgG Mouse Monoclonal Antibody
Anti-IgG Mouse Monoclonal Antibody
  BIRBORB43307-100
 :  Biorbyt
undefined
Anti-IgG Mouse Monoclonal Antibody
  BIRBORB43307-100
 :  Biorbyt
 
 
   
  1 * 100 µl

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Gel shift:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Goat
  • Western blot:
    Yes
  • Storage buffer:
    TBS (pH 7,4), 0,5% BSA, 25% Glycerol. Preservative: 0,05% Sodium Azide
  • Molecular weight:
    25 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    The immunogen used is goat IgG.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Goat