Northern Ireland
Order Entry
Anti-BCL2 Rabbit polyclonal antibody
  BIRBORB43279-100
 :  Biorbyt
Anti-BCL2 Rabbit polyclonal antibody
  BIRBORB43279-100
 :  Biorbyt
 
 
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    B-cell CLL/lymphoma 2
  • Antigen symbol:
    BCL2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Amino acid number:
    16 to 29
  • Storage buffer:
    TBS (pH 7,4), 0,5% BSA, 25% Glycerol. Preservative: 0,05% Sodium Azide
  • Molecular weight:
    26 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    The immunogen used is Bcl-2(aa16-29).
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: BCL2 (B-cell CLL/lymphoma 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat