Anti-BCL2L10 Rabbit Polyclonal Antibody
Catalog # BIRBORB41707-100
Supplier: Biorbyt
- Antibody type:Primary
- Antigen name:BCL2-like 10 (apoptosis facilitator)
- Antigen symbol:BCL2L10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:70 to 133
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide contain a sequence corresponding to a region within amino acids 70 and 133 of BCL2L10.
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250
Type: Primary
Antigen: BCL2L10 (BCL2-like 10 (apoptosis facilitator))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human