Northern Ireland
Order Entry
Anti-LITAF Rabbit Polyclonal Antibody
Anti-LITAF Rabbit Polyclonal Antibody
  BIRBORB41515-100
 :  Biorbyt
Anti-LITAF Rabbit Polyclonal Antibody
  BIRBORB41515-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Lipopolysaccharide-induced TNF Factor
  • Antigen symbol:
    LITAF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Amino acid number:
    1 to 132
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 132 of Human LITAF.
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Western Blot: 1:500-1:3000

Type: Primary
Antigen: LITAF (lipopolysaccharide-induced TNF factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human