Northern Ireland
Order Entry
Anti-KRR1 Rabbit Polyclonal Antibody
Anti-KRR1 Rabbit Polyclonal Antibody
Catalog # BIRBORB41188-100
Supplier:  Biorbyt
Anti-KRR1 Rabbit Polyclonal Antibody
  BIRBORB41188-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    KRR1, small subunit (SSU) processome component, homolog (yeast)
  • Antigen symbol:
    KRR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Amino acid number:
    47 to 314
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant protein fragment contain a sequence corresponding to a region within amino acids 47 and 314 of KRR1.
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: KRR1 (KRR1, small subunit (SSU) processome component, homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human