Anti-DCTN1 Rabbit Polyclonal Antibody
BIRBORB40761-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Dynactin 1
- Antigen symbol:DCTN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:1216 to 1278
- Storage buffer:1XPBS, 1%BSA, 20% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide contain a sequence corresponding to a region within amino acids 1216 and 1278 of dynactin 1.
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: DCTN1 (dynactin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human