Anti-IgG Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
: Biorbyt
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
- Catalog No:
- BIRBORB27730-0.5
- BIRBORB27730-1
- Antigen symbol:
- IgG
- IgG
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- TRITC (Tetramethylrhodamine Isothiocyanate)
- TRITC (Tetramethylrhodamine Isothiocyanate)
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- SizePk
Specifications
Antigen symbolIgGEnvironmentally PreferableAntibody typeSecondaryClonalityPolyclonalConjugationTRITC (Tetramethylrhodamine Isothiocyanate)ReactivityRabbitHostGoatIsotypeIgGWestern blotYesELISAYesImmunoChemistryYesImmunoFluorescenceYesStorage temperatureCoolShipping temperatureIceSpecifications
Antigen symbolIgGEnvironmentally PreferableAntibody typeSecondaryClonalityPolyclonalConjugationTRITC (Tetramethylrhodamine Isothiocyanate)ReactivityRabbitHostGoatIsotypeIgGWestern blotYesELISAYesImmunoChemistryYesImmunoFluorescenceYesStorage temperatureCoolShipping temperatureIce