Northern Ireland
Order Entry
Anti-IgG Rabbit Antibody
Anti-IgG Rabbit Antibody
  BIRBORB27706-0.5
 :  Biorbyt
Anti-IgG Rabbit Antibody
Catalog # BIRBORB27706-0.5
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 0,5 mg

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Goat
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 μg
  • Pk:
    0,5 mg

Specifications

About this item

Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat