Anti-Caspase-1 P20 Rabbit Polyclonal Antibody
Catalog # BIRBORB27641-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Caspase 1 P20
- Antigen symbol:CASP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Caspase-1 P20
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human