Northern Ireland
Order Entry
Anti-Multilink Horse Polyclonal Antibody (Biotin)
  BIRBORB27529-1
 :  Biorbyt
Anti-Multilink Horse Polyclonal Antibody (Biotin)
  BIRBORB27529-1
 :  Biorbyt
 
 
   
  1 * 1 mg

 

  • Antibody type:
    Secondary
  • Antigen name:
    Multilink
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Horse
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Format:
    concentrated
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Concentration:
    dilute 1:50 up to 1:250
  • Shipping temperature:
    Ice
  • Immunogen:
    Affinity purified IgG from mouse, rabbit and goat
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Secondary
Antigen: Multilink
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Horse
Isotype: none
Reactivity: