Anti-PSAT1 Mouse Monoclonal Antibody
BIRBORB27460-5
: Biorbyt
- Antibody type:Primary
- Antigen name:Phosphoserine Aminotransferase 1
- Antigen symbol:PSAT1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Concentration:neat
- Shipping temperature:Ice
- Immunogen:Purified human prostate-specific antigen
- Size:5 ml
- Pk:5 mL
Type: Primary
Antigen: PSAT1 (phosphoserine aminotransferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human