Anti-NCAM1 Mouse Monoclonal Antibody
BIRBORB27423-3
: Biorbyt
- Antibody type:Primary
- Antigen name:Neural Cell Adhesion Molecule 1
- Antigen symbol:NCAM1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Storage temperature:Cool
- Concentration:neat
- Shipping temperature:Ice
- Immunogen:Preparation of a SCLC-derived cell line
- Size:3 ml
- Pk:3 mL
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: