Anti-COL4 Mouse Monoclonal Antibody
BIRBORB27341-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:collagen, type IV
- Antigen symbol:COL4
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Purified collagen type IV from human placenta
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Immunochemistry: > 1:4000; ELISA: >1:4000
Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human