Northern Ireland
Order Entry
Anti-COL2 Rabbit Polyclonal Antibody
Anti-COL2 Rabbit Polyclonal Antibody
 :  Biorbyt
 
Anti-COL2 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB27339-0.1
  • BIRBORB27339-0.5
  • Antigen symbol:
  • COL2
  • COL2
  • Antigen name:
  • collagen, type II
  • collagen, type II
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • Purified collagen type II from fetal murine cartilage
  • Purified collagen type II from fetal murine cartilage
  • Form:
  • lyophilized
  • lyophilized
  • Purification:
  • purified
  • purified
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

  •  
     
    Size
    Pk
     
     
  • BIRBORB27339-0.1
    ORB27339-0.1
    100 μg
    0,1 mg
       
       
    1 * 0,1 mg
    1 * 0,1 mg

     
     
    Specifications
    Antigen symbol
    COL2
    Antigen name
    collagen, type II
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Immunogen
    Purified collagen type II from fetal murine cartilage
    Form
    lyophilized
    Purification
    purified
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB27339-0.5
    ORB27339-0.5
    500 μg
    0,5 mg
       
       
    1 * 0,5 mg
    1 * 0,5 mg

     
     
    Specifications
    Antigen symbol
    COL2
    Antigen name
    collagen, type II
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Immunogen
    Purified collagen type II from fetal murine cartilage
    Form
    lyophilized
    Purification
    purified
    Storage temperature
    Cool
    Shipping temperature
    Ice