Anti-KRT17 Mouse Monoclonal Antibody
BIRBORB27189-1
: Biorbyt
- Antibody type:Primary
- Antigen name:keratin 17
- Antigen symbol:KRT17
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Format:concentrated
- Form:liquid
- Storage temperature:Cool
- Concentration:1:10 - 1:50
- Shipping temperature:Ice
- Immunogen:Human Cytokeratin 17 , MW 46 kDa
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: KRT17 (keratin 17)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: