Anti-Laminin Rabbit Polyclonal Antibody
BIRBORB26852-0.5
: Biorbyt
- Antibody type:Primary
- Antigen name:Laminin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Mouse Tumor Tissues
- Purification:purified by chromatography
- Size:500 μL
- Pk:0,5 mL
- ELISA: 1:200; Immunofluorescence: 1:80; Immunochemistry: 1:500
Type: Primary
Antigen: Laminin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse