Anti-COL4 Rabbit Polyclonal Antibody
BIRBORB26851-0.5
: Biorbyt
- Antibody type:Primary
- Antigen name:collagen, type IV
- Antigen symbol:COL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Collagen Type IV extracted and purified from mouse tumor tissues.
- Purification:purified by chromatography
- Size:500 μL
- Pk:0,5 mL
Immunofluorescence: 1:80; Immunochemistry: 1:500; ELISA: 1:200
Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse