Northern Ireland
Order Entry
Anti-Staph. Enterotoxin D Rabbit Polyclonal Antibody
  BIRBORB26798-1
 :  Biorbyt
Anti-Staph. Enterotoxin D Rabbit Polyclonal Antibody
  BIRBORB26798-1
 :  Biorbyt
 
 
   
  1 * 1 mg

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Staph. Enterotoxin D
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Form:
    lyophilized
  • Storage buffer:
    0,01M phosphate, pH 7,4 containing 0,9% Sodium chloride
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml (prior to lyophilization)
  • Shipping temperature:
    Ice
  • Immunogen:
    Staphylococcus aureus Enterotoxin D (SED)
  • Purification:
    purified by Toxin specific immunoaffinity column
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: Staph. Enterotoxin D
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: