Northern Ireland
Order Entry
Anti-Cortisol Rabbit Polyclonal Antibody
  BIRBORB26782-1
 :  Biorbyt
Anti-Cortisol Rabbit Polyclonal Antibody
  BIRBORB26782-1
 :  Biorbyt
 
 
   
  1 * 1 mL

 

  • Antibody type:
    Primary
  • Antigen name:
    Cortisol
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Primary
Antigen: Cortisol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human