Northern Ireland
Order Entry
Anti-APP Mouse monoclonal antibody
Anti-APP Mouse monoclonal antibody
  BIRBORB26763-100
 :  Biorbyt
undefined
Anti-APP Mouse monoclonal antibody
  BIRBORB26763-100
 :  Biorbyt
 
 
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid Beta (A4) Precursor Protein
  • Antigen symbol:
    APP
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Form:
    lyophilized
  • Antigen synonyms:
    ABETA|beta Amyloid
  • Amino acid number:
    1 to 42
  • Storage buffer:
    2X PBS, PEG and Sucrose
  • Storage temperature:
    Cool
  • Concentration:
    100 ug/ml (prior to lyophilization)
  • Shipping temperature:
    Ice
  • Immunogen:
    C-terminal peptide conjugated to KLH.
  • Purification:
    purified by Thiophilic adsorption and size exclusion chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human