Northern Ireland
Order Entry
Anti-APP Mouse monoclonal antibody
  BIRBORB26648-0.2
 :  Biorbyt
Anti-APP Mouse monoclonal antibody
  BIRBORB26648-0.2
 :  Biorbyt
 
 
   
  1 * 0,2 mL

 

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid Beta (A4) Precursor Protein
  • Antigen symbol:
    APP
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    ABETA|beta Amyloid
  • Amino acid number:
    1 to 42
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml (OD280)
  • Shipping temperature:
    Ice
  • Immunogen:
    Peptide corresponding to the last 6 amino acids of ABx-42 coupled to KLH through the addition of an N-terminal cysteine.
  • Purification:
    purified by Protein G chromatography
  • Size:
    200 μL
  • Pk:
    0,2 mL

 

 

Western Blot: 1:100-1:1000; ELISA: 1:10000-1:1000000; Immunochemistry: 1:100-1:2000

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human