Anti-Mouse CD28 Hamster Monoclonal Antibody
Catalog # BIRBORB26397-0.5
Supplier: Biorbyt
- Antibody type:Primary
- Antigen symbol:CD28
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Storage buffer:0,01M PBS, pH 7,2 containing 1% BSA.
- Storage temperature:Cool
- Concentration:0,5 mg/ml (OD280)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:500 μg
- Pk:0,5 mg
About this item
Flow Cytometry: 1 ?g
Type: Primary
Antigen: Mouse CD28
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG (Hamster)
Reactivity: Mouse