Northern Ireland
Order Entry
Anti-FUT4 Mouse Monoclonal Antibody
Catalog # BIRBORB26225-100
Supplier:  Biorbyt
Anti-FUT4 Mouse Monoclonal Antibody
Catalog # BIRBORB26225-100
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Fucosyltransferase 4
  • Antigen symbol:
    FUT4
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Human Granulocytes
  • Purification:
    purified by precipitation methods and ion exchange chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

About this item

Type: Primary
Antigen: FUT4 (fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human