Anti-Cortisol Mouse Monoclonal Antibody
BIRBORB26162-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Cortisol
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Storage temperature:Cool
- Concentration:5,59 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Purification:purified
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Cortisol
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human