Anti-CEACAM5 Mouse monoclonal antibody
BIRBORB26141-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Carcinoembryonic antigen-related cell adhesion molecule 5
- Antigen symbol:CEACAM5
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Storage buffer:10mM Sodium phosphate, pH 7,4 containing 150mM Sodium chloride
- Storage temperature:Cool
- Concentration:5,40 mg/ml (OD280, E(0,1%) = 1,4)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: CEACAM5 (carcinoembryonic antigen-related cell adhesion molecule 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human