Anti-ICAM1 Mouse Monoclonal Antibody
BIRBORB26128-0.2
: Biorbyt
- Antibody type:Primary
- Antigen name:intercellular adhesion molecule 1
- Antigen symbol:ICAM1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Storage buffer:10mM Sodium phosphate, pH 7,4 containing 150mM Sodium Chloride
- Storage temperature:Cool
- Concentration:1,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 μg
- Pk:0,2 mg
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human