Anti-hGH Mouse Monoclonal Antibody
Catalog # BIRBORB26076-1
Supplier: Biorbyt
- Antibody type:Primary
- Antigen symbol:HGH
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Storage buffer:10mM Phosphate, pH 7,4 containing 150mM Sodium chloride
- Storage temperature:Cool
- Concentration:5,04 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:hGH isolated from human pituitary gland.
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: hGH
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human