Northern Ireland
Order Entry
Anti-CKM Mouse monoclonal antibody
Anti-CKM Mouse monoclonal antibody
  BIRBORB26048-1
 :  Biorbyt
Anti-CKM Mouse monoclonal antibody
  BIRBORB26048-1
 :  Biorbyt
 
 
   
  1 * 1 mg

 

  • Antibody type:
    Primary
  • Antigen name:
    Creatine Kinase, muscle
  • Antigen symbol:
    CKM
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Storage buffer:
    10 mM Phosphate, pH 7,4 containing 150 mM Sodium Chloride.
  • Storage temperature:
    Cool
  • Concentration:
    5,2 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    CK-MM purified from human smooth muscle.
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: CKM (creatine kinase, muscle)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human