Northern Ireland
Order Entry
Anti-CRP Mouse monoclonal antibody
  BIRBORB25859-1
 :  Biorbyt
undefined
Anti-CRP Mouse monoclonal antibody
  BIRBORB25859-1
 :  Biorbyt
Price
Inquire For Price
Log in for availability
  1 * 1 mg

 

  • Antibody type:
    Primary
  • Antigen name:
    C-Reactive Protein
  • Antigen symbol:
    CRP
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    4,8 mg/ml (Sigma Protein Assay Kit)
  • Shipping temperature:
    Ice
  • Immunogen:
    Human CRP, derived from pleural/ascetic fluid or plasma.
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: CRP (C-reactive protein, pentraxin-related)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human